Order Entry
Singapore
ContactUsLinkComponent
Description: Haze-Gard Plus Illuminant C, Lab Instrument to objectively evaluate total appearance of transparent products Power supply: 115V/ 60 Hz (230V/ 50 Hz) Details for Reference Principle is 0 illumination, diffuse viewing geometry. Transpa 1 * 1 items
Catalog # IBSAGL-4725
Supplier:  IBS Stock Items
Description: Haze-Gard Plus Illuminant C, Lab Instrument to objectively evaluate total appearance of transparent products Power supply: 115V/ 60 Hz (230V/ 50 Hz) Details for Reference Principle is 0 illumination, diffuse viewing geometry. Transpa 1 * 1 items
Catalog # IBSAGL-4725
Supplier:  IBS Stock Items