Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Puradisc FP 30mm Syringe Filter, Cellulose Acetate, 0.2µm, sterile 10 pack (5x10462200) 1 * 1 SET
Catalog # WHAT514-1111BOM10
Supplier:
Whatman products (Cytiva)
Expand image
Puradisc FP 30mm Syringe Filter, Cellulose Acetate, 0.2µm, sterile 10 pack (5x10462200) 1 * 1 SET
Catalog # WHAT514-1111BOM10
Supplier:
Whatman products (Cytiva)