Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
BUFFER PHOSPHATE 0.2M PH 7.4 SORENSENS 1 * 4 l
Catalog # ELMI11601-40
Supplier:
ELECTRON MICROSCOPY SCIENCES
Expand image
BUFFER PHOSPHATE 0.2M PH 7.4 SORENSENS 1 * 4 l
Catalog # ELMI11601-40
Supplier:
ELECTRON MICROSCOPY SCIENCES