Digital Solutions
Applications
Support
About
Singapore
ContactUsLinkComponent
STYLE 26: VERY STRONG, SHARP TIPS LENGTH: 5 1/4 (135MM) 1 * 1 items
ELMI78328-26
:
ELECTRON MICROSCOPY SCIENCES
STYLE 26: VERY STRONG, SHARP TIPS LENGTH: 5 1/4 (135MM) 1 * 1 items
ELMI78328-26
:
ELECTRON MICROSCOPY SCIENCES