Digital Solutions
Applications
Support
About
Singapore
ContactUsLinkComponent
Serological Pipet, Glass, 5 ml, pk/1, Subdivisions 0.1 ml, Tolerance ± 0.04 ml, 1 * 1 items
HACH53237
:
Hach
Serological Pipet, Glass, 5 ml, pk/1, Subdivisions 0.1 ml, Tolerance ± 0.04 ml, 1 * 1 items
HACH53237
:
Hach