MOBS, 0,2 M buffer soln., pH 7,5
MOBS buffer solution, 0,2 M, 7,5, 250 mL
Supplier: Thermo Fisher Scientific
MOBS, 0,2 M buffer soln., pH 7,5
Catalog # ALFAJ63519.AK
Supplier: Thermo Fisher Scientific
CAS Number:
Jump to:
Product Details & Documents
CAS No.: 115724-21-5
Physical Form: Liquid
MDL No.: MFCD00674916
Documents
Specifications
Specifications
- Concentration:0,2 M
- Pk:250 mL
- Description:MOBS buffer solution
- pH value:7,5
- Cat. No.:ALFAJ63519.AK
- Physical form:Liquid
- MDL No.:MFCD00674916
- CAS No.:115724-21-5



