Buffers, PIPES, 1,0 M solutions
PIPES buffer solution, 1,0 mol/L, 7,0, Glass bottle, 250 mL
Supplier: Thermo Fisher Scientific
Buffers, PIPES, 1,0 M solutions
Catalog # ALFAJ62195.AK
Supplier: Thermo Fisher Scientific
CAS Number:
Jump to:
Product Details & Documents
Liquid
CAS No.: 5625-37-6
Appearance: Colorless
MDL No.: MFCD00006159
: Storage Temperature: 2 to 8 °C
Specifications
Specifications
- Concentration:1,0 mol/L
- Pack type:Glass bottle
- Pk:250 mL
- Description:PIPES buffer solution
- pH value:7,0
- Cat. No.:ALFAJ62195.AK
- Storage temperature:2°C to 8°C
- Physical form:Liquid
- MDL No.:MFCD00006159
- Appearance:Colourless
- CAS No.:5625-37-6



