EPPS, 0,2 M buffer solutions
EPPS buffer solution, 0,2 mol/L, 7,5, Glass bottle, 250 mL
CAS No.: 16052-06-5
Physical form: liquid
MDL No.: MFCD00006160
- Concentration:0,2 mol/L
- Pack type:Glass bottle
- Pk:250 mL
- Description:EPPS buffer solution
- pH value:7,5
- Cat. No.:ALFAJ60511.AK
- Physical form:Liquid
- MDL No.:MFCD00006160
- CAS No.:16052-06-5



