CHES, 0.5 M buffer solutions
CHES buffer solution, 0,5 M, 8,5, 250 mL
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
2-(Cyclohexylamino)ethanesulfonic acid, CHES.
CAS No.: 103-47-9
MDL No.: MFCD00003835
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Concentration:0,5 M
- Pk:250 mL
- Description:CHES buffer solution
- pH value:8,5
- Cat. No.:ALFAJ61492.AK
- Physical form:Liquid
- MDL No.:MFCD00003835
- CAS No.:103-47-9



