Order Entry
Singapore
ContactUsLinkComponent
Plug screwed, bore size 8.7mm, PTFE, for use with Torion glass bodies and rubber/PTFE sealing rings, fittings available in choice of materials, PTFE for use at temperatures up to 250C. 1 * 10 items
Catalog # BIBB720-05
Supplier:  SciLabware
Plug screwed, bore size 8.7mm, PTFE, for use with Torion glass bodies and rubber/PTFE sealing rings, fittings available in choice of materials, PTFE for use at temperatures up to 250C. 1 * 10 items
Catalog # BIBB720-05
Supplier:  SciLabware