Safety cabinet with wing doors for the storage of pressurised gas cylinders. Outer dimensions: 1195 x 596 x 2070 (WxDxH) mm. Inner d imension: 1100 x 439 x 1884 (WxDxH) mm. weight: approx. 505 kg Outer carcass in sheet steel powder coated in RAL 7035 1 * 1 items
Safety cabinet with wing doors for the storage of pressurised gas cylinders. Outer dimensions: 1195 x 596 x 2070 (WxDxH) mm. Inner d imension: 1100 x 439 x 1884 (WxDxH) mm. weight: approx. 505 kg Outer carcass in sheet steel powder coated in RAL 7035 1 * 1 items