Order Entry
Singapore
ContactUsLinkComponent
Put-in tube extension, outer diam. 60,3 mm, inner 57 mm, angled, for exhaust air ducting, if necessary for connection by hose, for u n 1 * 1 items
Catalog # MEMMB29719UN110
Supplier:  MEMMERT
Put-in tube extension, outer diam. 60,3 mm, inner 57 mm, angled, for exhaust air ducting, if necessary for connection by hose, for u n 1 * 1 items
Catalog # MEMMB29719UN110
Supplier:  MEMMERT