Digital Solutions
Applications
Support
About
Singapore
ContactUsLinkComponent
Put-in tube extension, outer diam. 60,3 mm, inner 57 mm, angled, for exhaust air ducting, if necessary for connection by hose, for u fplus 1 * 1 items
MEMMB29719UF750PLS
:
MEMMERT
Put-in tube extension, outer diam. 60,3 mm, inner 57 mm, angled, for exhaust air ducting, if necessary for connection by hose, for u fplus 1 * 1 items
MEMMB29719UF750PLS
:
MEMMERT