Order Entry
Singapore
Orders LinkContactUsLinkComponent
BASIC Plus Laboratory Coats
BASIC Plus Laboratory Coats
White, S
Catalog # KIMW10020
   Waste Reducing
Supplier:   KIMBERLY CLARK
BASIC Plus Laboratory Coats
Catalog # KIMW10020
Supplier:  KIMBERLY CLARK
   Waste Reducing
BASIC Plus Laboratory Coats
BASIC Plus Laboratory Coats
White, S
Price
Inquire For Price
Log in for availability
1 * 25 items
Product Details & Documents
These coats are made of a protective three-layer SMS fabric which is resistant to blood strike-through. Recommended for areas where there is the potential for light fluid contact.
  • Knit collar and cuffs
  • Five snap buttons
  • Three reinforced pockets
  • With size/name label
  • Neck loop for hanging
Specifications
  • Clothing Size:
    S
  • Colour:
    White
  • Seq:
    6
  • Cat. No.:
    KIMW10020
  • Garment application:
    Laboratory|Clinical
  • Gender:
    Unisex
  • Pockets:
    With pockets
  • Closure type:
    Press studs / snaps
  • Environmentally Preferable:
    Waste Reducing