D-(-)-Fructose ≥99%, high purity
Plastic bucket, 12 kg
: D(-)-Levulose, (3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one, β-D-(-)-Fructose, Fructose
- :57-48-7
- :C₆H₁₂O₆
- :MFCD00148910
- :180.16 g/mol
- :13,04295
- :552 °C (1013 hPa)
- :100…110 °C
- :1,59 g/cm³ (20 °C. OECD 109)
- :Ambient
- :200-333-3
- Arsenic:<= 0.0001%
- Calcium & Magnesium:<= 0.005%
- Chloride:<= 0.018%
- Heavy Metals (as Pb):<= 0.0005%
- Loss on Drying:<= 0.5%
- Purity:>= 99%
- Residue on Ignition:<= 0.5%
- Sulphate:<= 0.025%
- Pack type:Plastic bucket
- Pk:12 kg



