Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
ETO indicator dot 8.01.17, with text Green is ETO Exposed Dia 12.5 (5,000pcs/roll) 1 * 1 Roll
Catalog # XXXX8.01.17
Supplier:
Chemicals
Expand image
ETO indicator dot 8.01.17, with text Green is ETO Exposed Dia 12.5 (5,000pcs/roll) 1 * 1 Roll
Catalog # XXXX8.01.17
Supplier:
Chemicals