$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Singapore
ContactUsLinkComponent
Recombinant protein, gpr175( np_057456, 281 a.a. - 371 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
ABNOH00131601-Q01
:
Abnova
Recombinant protein, gpr175( np_057456, 281 a.a. - 371 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
ABNOH00131601-Q01
:
Abnova
sds_ABNOH00131601-Q01.pdf
TDS_ABNOH00131601-Q01.pdf