Order Entry
Singapore
ContactUsLinkComponent
Recombinant protein, otx1( np_055377.1, 1 a.a. - 354 a.a.) full-length recombinant protein with gst tag. 1 * 2 µG
Catalog # ABNOH00005013-P01
Supplier:  Abnova
Recombinant protein, otx1( np_055377.1, 1 a.a. - 354 a.a.) full-length recombinant protein with gst tag. 1 * 2 µG
Catalog # ABNOH00005013-P01
Supplier:  Abnova
Related Information