$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Singapore
ContactUsLinkComponent
Recombinant protein, orc2l( aah14834, 1 a.a. - 578 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
ABNOH00004999-P01
:
Abnova
Recombinant protein, orc2l( aah14834, 1 a.a. - 578 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
ABNOH00004999-P01
:
Abnova