$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Singapore
ContactUsLinkComponent
Recombinant protein, cryz( np_001880.2, 1 a.a. - 329 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
ABNOH00001429-P01
:
Abnova
Recombinant protein, cryz( np_001880.2, 1 a.a. - 329 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
ABNOH00001429-P01
:
Abnova
sds_ABNOH00001429-P01.pdf
TDS_ABNOH00001429-P01.pdf