Biospectrophotometers, BioSpectrometer® basic
UV/Vis biospectrophotometer, EU-plug, BioSpectrometer® basic, 200 − 830 nm
Catalog # EPPE6135000.009
Jump to:
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
BioSpectrometer basic UK-plug 230V/50-60Hz 100-240V UV/Vis spectrophotometer wavelengths 200nm to 830nm 1 * 1 items Catalog #EPPE6135000.025
Product EPPE6135000.025 is a replacement for EPPE6135000.009 and has the same packaging and specifications, but may be a different quantity.
Product Details & Documents
The BioSpectrometer basic are designed for measurements in the UV and visible range.
- Xenon flash lamp with very long service life
- Guided software process to minimise errors
- Automatic evaluation and storage of measurement data with clear presentation of results
- Integrated data processing option for specific methods
- Two-wavelength method with substraction or division evaluation
- Integrated application and results memory, self-test and calibration history
Spectra can be recorded and wavelength measurements can be made.
Applications include nucleic acid quantification and direct protein quantification, microvolume measurement, bacteria growth, colorimetric assays, cytotoxicity assays.
Delivery information: Supplied with power cable and multi-language manual.
Specifications
- Model:BioSpectrometer® basic
- Range:200 − 830 nm
- Description:UV/Vis biospectrophotometer, EU-plug
- Cat. No.:EPPE6135000.009
- Light source:Pulsed xenon
- Detector:CMOS photodiode array
- Optical design:Double beam
- Memory:Over 1000 values
- W×D×H:295×400×150 mm
- Measuring modes:Extinction measurements with one or several wavelengths, wavelength scans ; OD600 dye methods ; FOI ; analysis via factor
- Weight:5,4 kg
- Absorbance range:0 − 3 A
- Display:(VGA) TFT LCD 145 mm (8")
- Spectral bandwidth:<4 nm



