Labels, hazard warning
Hazard labels: low temperature, PIC 323-TRI 200-B7541, Polyester, Black on yellow, 200×173 mm, 1 label/card
Catalog # BRDY250271
Supplier: Brady
Jump to:
Product Details & Documents
These self adhesive signs are designed to warn people to be careful and to take precautions of potential hazards.
Available in different sizes.
: Conforms to Health and Safety (Safety Signs and Signals) Regulations 1996.
Specifications
- Colour:Black on yellow
- Description:Hazard labels: low temperature
- Dimensions:200×173 mm
- Material:Polyester
- Seq:42
- Model:PIC 323-TRI 200-B7541
- Packed:1 label/card