Labels, hazard warning
Hazard labels: handling vehicle, —, Polyester, Black on yellow, 200×173 mm, 1 label/card
BRDY250220
: Brady
These self adhesive signs are designed to warn people to be careful and to take precautions of potential hazards.
Available in different sizes.
: Conforms to Health and Safety (Safety Signs and Signals) Regulations 1996.
- Colour:Black on yellow
- Description:Hazard labels: handling vehicle
- Dimensions:200×173 mm
- Material:Polyester
- Seq:35
- Model:—
- Packed:1 label/card