Labels, hazard warning
Hazard labels: conductive shoes, —, Polyester, Black on yellow, 200×173 mm, 250 labels/roll
Catalog # BRDY250144
Supplier: Brady
Jump to:
Product Details & Documents
These self adhesive signs are designed to warn people to be careful and to take precautions of potential hazards.
Available in different sizes.
Certifications: Conforms to Health and Safety (Safety Signs and Signals) Regulations 1996.
Specifications
- Colour:Black on yellow
- Description:Hazard labels: conductive shoes
- Dimensions:200×173 mm
- Material:Polyester
- Seq:33
- Model:—
- Packed:250 labels/roll



