Order Entry
Singapore
Orders LinkContactUsLinkComponent
Labels, hazard warning
Hazard labels: strong magnetic field, PIC 322-TRI 200-B7541, Polyester, Black on yellow, 200×173 mm, 1 label/card
  BRDY250268
 :   Brady
Labels, hazard warning
  BRDY250268
 :  Brady
Labels, hazard warning
Labels, hazard warning
Hazard labels: strong magnetic field, PIC 322-TRI 200-B7541, Polyester, Black on yellow, 200×173 mm, 1 label/card
$12.90
1 * 1 items
These self adhesive signs are designed to warn people to be careful and to take precautions of potential hazards.
Available in different sizes.
 : Conforms to Health and Safety (Safety Signs and Signals) Regulations 1996.
  • Colour:
    Black on yellow
  • Description:
    Hazard labels: strong magnetic field
  • Dimensions:
    200×173 mm
  • Material:
    Polyester
  • Seq:
    41
  • Model:
    PIC 322-TRI 200-B7541
  • Packed:
    1 label/card