Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
BUFFER PH 11.00 0.01, MEETS USP REQ 1 * 500 mL
Catalog # RICC1610-16
Supplier:
Ricca Chemical
Expand image
BUFFER PH 11.00 0.01, MEETS USP REQ 1 * 500 mL
Catalog # RICC1610-16
Supplier:
Ricca Chemical