Stoppers
Yellow, 19/22 - 19/38
CGLACG-3021-02
Marketsource
These tapered stopper provides for a tight, leakproof ft while the top ring permits the stopper to be easily removed.
- PE
- Yellow
- NS:19/22 - 19/38
- Colour:Yellow
- Seq:01
These tapered stopper provides for a tight, leakproof ft while the top ring permits the stopper to be easily removed.