Order Entry
Singapore
Orders LinkContactUsLinkComponent
VWR®, Accessories for Circulators
VWR®, Accessories for Circulators
PT100 probe with 7,6 m cable
Catalog # VWRI462-0273
Supplier:   VWR Collection
VWR®, Accessories for Circulators
Catalog # VWRI462-0273
Supplier:  VWR Collection
VWR®, Accessories for Circulators
VWR®, Accessories for Circulators
PT100 probe with 7,6 m cable
Price
$1,960.00
Log in for availability
1 * 1 items
Product Details & Documents
Stainless steel probes suitable for used with any instrument that has an Advanced Digital (AD) or Advanced Programmable (AP) controller.
Probe measurements: 152 mm length, 4,7 mm Ø.
Specifications
  • Description:
    PT100 probe with 7,6 m cable