Order Entry
Singapore
Orders LinkContactUsLinkComponent
Evaporating flasks
Evaporating flasks
200 ml, 24/40 NS
  CGLACG-1512-05
Marketsource
 :   CHEMGLASS
Evaporating flasks
  CGLACG-1512-05
 :  CHEMGLASS
Marketsource
Evaporating flasks
Evaporating flasks
200 ml, 24/40 NS
$303.00
1 * 1 items
Made of borosilicate glass, coated.
  • Flasks with 24/40 joints have a square reniforcing bead
  • Plastic coated to prevent injury from broken glass, and also prevents spillage of hazardous materials
  • Very good chemical resistance
These single neck recovery flask can be used as an evaporating flask in rotary evaporators.
The flask have a heavy wall for vaccum applications and are balance to prevent run-out during rotation.
  • Socket size:
    24/40 NS
  • Capacity:
    200 ml
  • Seq:
    06