Order Entry
Singapore
Orders LinkContactUsLinkComponent
Evaporating flasks
Evaporating flasks
100 ml, 24/40 NS
Catalog # CGLACG-1512-03
Marketsource
Supplier:   CHEMGLASS
Evaporating flasks
Catalog # CGLACG-1512-03
Supplier:  CHEMGLASS
Marketsource
Evaporating flasks
Evaporating flasks
100 ml, 24/40 NS
Price
$231.00
Log in for availability
1 * 1 items
Product Details & Documents
Made of borosilicate glass, coated.
  • Flasks with 24/40 joints have a square reniforcing bead
  • Plastic coated to prevent injury from broken glass, and also prevents spillage of hazardous materials
  • Very good chemical resistance
These single neck recovery flask can be used as an evaporating flask in rotary evaporators.
The flask have a heavy wall for vaccum applications and are balance to prevent run-out during rotation.
Specifications
  • Socket size:
    24/40 NS
  • Capacity:
    100 ml
  • Seq:
    05