Büchner funnels
Cone NS 24/40, 60 mm, 150 ml, Medium (10 - 16 µm)
Catalog # CGLACG-1406-11
Marketsource
Supplier: CHEMGLASS
Jump to:
Product Details & Documents
Büchner style flter funnels with a lower standard taper inner joint and serrated tubulation for connection to a vacuum source.
- No filter papers required
- Highly resistant to most corrosive reagents including ammonia and concentrated sulphuric acid
- Very good temperature resistance
For convenient filtration of solutions.
Specifications
- Plate Ø:60 mm
- Porosity:Medium (10 - 16 µm)
- Capacity:150 ml
- Description:Cone NS 24/40
- Seq:01



