Order Entry
Singapore
ContactUsLinkComponent
Temperature controller, safety device, class3.3(DIN 12880) with visual alarm, 230V1N50/60Hz for KMF 115 (E5.2) series 1 * 1 items
Catalog # BIND8012-0703
Supplier:  Binder
Temperature controller, safety device, class3.3(DIN 12880) with visual alarm, 230V1N50/60Hz for KMF 115 (E5.2) series 1 * 1 items
Catalog # BIND8012-0703
Supplier:  Binder