Order Entry
Singapore
ContactUsLinkComponent
Improved design permits greater condensing capacity with up to double the cycling rate of conventional-style extractors. All extractors feature a [ST] 24/40 bottom joint. 1 * 1 items
  ACEG6716-16
 :  Ace Glass
Improved design permits greater condensing capacity with up to double the cycling rate of conventional-style extractors. All extractors feature a [ST] 24/40 bottom joint. 1 * 1 items
  ACEG6716-16
 :  Ace Glass