Order Entry
Singapore
ContactUsLinkComponent
Gloves Starpack :Ambidextrous Nitrile. Powderfree. Textured Fingertips. Thickness 0.08 mm (3 mil). Length 240 mm (9 inch). Blue.Size 8.5-9 1 * 200 items
Catalog # ANSE48-651/8.5-9
Supplier:  Ansell
Gloves Starpack :Ambidextrous Nitrile. Powderfree. Textured Fingertips. Thickness 0.08 mm (3 mil). Length 240 mm (9 inch). Blue.Size 8.5-9 1 * 200 items
Catalog # ANSE48-651/8.5-9
Supplier:  Ansell