Order Entry
Singapore
ContactUsLinkComponent
Black phenolic cap has a polyethylene cone liner, which molds itself around sealing areas of the finish as the cap is screwed on. Liner is secured to the protruding lock-stud in cap, to prevent dislodging or fall out. 1 * 12 items
  QORP5029/12
Black phenolic cap has a polyethylene cone liner, which molds itself around sealing areas of the finish as the cap is screwed on. Liner is secured to the protruding lock-stud in cap, to prevent dislodging or fall out. 1 * 12 items
  QORP5029/12