Order Entry
Singapore
ContactUsLinkComponent
GLOVES STARPACK AMBIDEXTROUS NITRILE. POWDERFREE. TEXTURED FINGERTIPS.THICKNESS 0.10 MM (4 MIL). L. 240 MM (9 INCH). WHITE.S.6,5-7 1 * 100 items
Catalog # ANSE48-601/6.5-7
Supplier:  Ansell
GLOVES STARPACK AMBIDEXTROUS NITRILE. POWDERFREE. TEXTURED FINGERTIPS.THICKNESS 0.10 MM (4 MIL). L. 240 MM (9 INCH). WHITE.S.6,5-7 1 * 100 items
Catalog # ANSE48-601/6.5-7
Supplier:  Ansell