Order Entry
Singapore
ContactUsLinkComponent
Digital Peristaltic Pump 230V, 50Hz. With head, console drive, and starter tubing kit (7.6m of 4.8mm O.D. general PVC tubing 4 hose clamps 2 each of 1.6mm, 3.2mm, and 4.8mm O.D. silicone tubing cartridges). Bi-directional speed control. Flow rate: 1 * 1 items
Catalog # GARD3200C-02
Supplier:  GARDNER DENVER
Digital Peristaltic Pump 230V, 50Hz. With head, console drive, and starter tubing kit (7.6m of 4.8mm O.D. general PVC tubing 4 hose clamps 2 each of 1.6mm, 3.2mm, and 4.8mm O.D. silicone tubing cartridges). Bi-directional speed control. Flow rate: 1 * 1 items
Catalog # GARD3200C-02
Supplier:  GARDNER DENVER