Order Entry
Singapore
Orders LinkContactUsLinkComponent
 

Distilling flasks, PYREX®

250 ml, 250 mm, 86 mm
  CORN4620-250
 :   Corning
Distilling flasks, PYREX®
  CORN4620-250
 :  Corning
Distilling flasks, PYREX®
Distilling flasks, PYREX®

250 ml, 250 mm, 86 mm

 
$54.20
   
1 * 1 items
 

PYREX® borosilicate glass.

  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes

The sidearm of the 125 ml flask is located approximately 137 mm from the bottom of the flask as required in ASTM E1405 and can be used in ASTM Tests D86 and D447. This size has a heavy-duty neck.

The sidearm on the 250 ml size is placed 77 mm below the top of the neck at a 75° angle and can be used with thermometers calibrated for 76 mm immersion.

 

 
  • Ext.Ø:
    86 mm
  • Capacity:
    250 ml
  • Height:
    250 mm
  • Seq:
    04