Order Entry
Singapore
Orders LinkContactUsLinkComponent
 

Volumetric pipettes, class A, serialised, PYREX®

3 ml, 0,01 ml ± ml, Black, 356 mm
Catalog # CORN7103C-3
Supplier:   Corning
Volumetric pipettes, class A, serialised, PYREX®
Catalog # CORN7103C-3
Supplier:  Corning
Volumetric pipettes, class A, serialised, PYREX®
Volumetric pipettes, class A, serialised, PYREX®

3 ml, 0,01 ml ± ml, Black, 356 mm

Price
$256.00
Log in for availability
1 * 6 items
Product Details & Documents

These PYREX® volumetric pipettes are calibrated to Class A tolerances in accordance with ASTM E-542 and ASTM E-969.

  • Reusable precision pipettes
  • Colour-coded markings are enameled onto the glass

PYREX® serialised/certified volumetric glassware is now tested and calibrated in an ISO/IEC 17025 certified laboratory.

Each pipette is individually serialized and supplied with a Certificate of Identification and capacity, traceable to NIST standards. These precision PYREX® volumetric pipettes are manufactured and calibrated 'To Contain' (TC: bottom line) and 'To Deliver' (TD: top line) to specifications for Class A volumetric ware.

 

Specifications
  • Colour code:
    Black
  • Capacity:
    3 ml
  • Length:
    356 mm
  • Seq:
    03
  • Tolerance:
    0,01 ml ± ml