Order Entry
Singapore
Orders LinkContactUsLinkComponent
Extraction thimbles, PYREX®
Extraction thimbles, PYREX®
25×85 mm, Coarse porosity fritted thimble, 40-60 μm
  CORN33950-SC
 :   Corning
Extraction thimbles, PYREX®
  CORN33950-SC
 :  Corning
Extraction thimbles, PYREX®
Extraction thimbles, PYREX®
25×85 mm, Coarse porosity fritted thimble, 40-60 μm
$867.00
1 * 9 items
PYREX®, borosilicate glass, with fritted disc.
  • Resistant to chemical attack
  • Excellent temperature resistance
It is designed for use with soxhlet extractors. It is also useful for chemical applications where it is required to weigh the extracted material.
Documents
  • Description:
    Coarse porosity fritted thimble, 40-60 μm
  • Seq:
    01
  • Ø ext.×H:
    25×85 mm