Extraction thimbles, PYREX®
45×130 mm, Extra coarse porosity fritted thimble, 170-220 μm
Catalog # CORN33950-LEC
Supplier: Corning
Jump to:
Product Details & Documents
PYREX®, borosilicate glass, with fritted disc.
- Resistant to chemical attack
- Excellent temperature resistance
It is designed for use with soxhlet extractors. It is also useful for chemical applications where it is required to weigh the extracted material.
Documents
Certificates
Search by Lot NumberSpecifications
- Description:Extra coarse porosity fritted thimble, 170-220 μm
- Seq:05
- Ø ext.×H:45×130 mm