Extraction thimbles, PYREX®
45×130 mm, Coarse porosity fritted thimble, 40-60 μm
Supplier: Corning
Jump to:
Product Details & Documents
PYREX®, borosilicate glass, with fritted disc.
- Resistant to chemical attack
- Excellent temperature resistance
It is designed for use with soxhlet extractors. It is also useful for chemical applications where it is required to weigh the extracted material.
Documents
Certificates
Search by Lot NumberSpecifications
- Description:Coarse porosity fritted thimble, 40-60 μm
- Seq:03
- Ø ext.×H:45×130 mm