Order Entry
Singapore
Orders LinkContactUsLinkComponent
Chromatography jar, rectangular, PYREX®
11,4 l, 238×181×324 mm
  CORN6944-11L
 :   Corning
Chromatography jar, rectangular, PYREX®
  CORN6944-11L
 :  Corning
Chromatography jar, rectangular, PYREX®
Chromatography jar, rectangular, PYREX®
11,4 l, 238×181×324 mm
$1,000.00
1 * 1 items
Jars are ground to the close tolerance needed for a tight cover fit.
  • These rectangular chromatography jars are made of Pyrex® borosilicate glass
Edges are ground flat within 0,25mm (0,01").
 : Covers are not supplied.
 : Not recommended for use with direct heat, pressure, or vacuum applications.
  • Capacity:
    11,4 l
  • W×D×H:
    238×181×324 mm