Order Entry
Singapore
Orders LinkContactUsLinkComponent
Drying trays, PYREX®

Drying trays, PYREX®

1600 ml, 349×249×60 mm
Catalog # CORN3175-8
Supplier:   Corning
Drying trays, PYREX®
Catalog # CORN3175-8
Supplier:  Corning
Drying trays, PYREX®
Drying trays, PYREX®

1600 ml, 349×249×60 mm

Price
$119.00
Log in for availability
1 * 4 items
Product Details & Documents

These dishes made of PYREX® borosilicate glass will withstand hot air or steam sterilization and are useful for drying, staining or developing trays.

  • Excellent chemical resistance
  • Good temperature resistance
  • Easy to clean

They can be used with gel electrophoresis or paper chromatography.

Caution: For oven use only. Do not use over direct heat.

Documents

Specifications
  • Capacity:
    1600 ml
  • Seq:
    02
  • W×D×H:
    349×249×60 mm