Order Entry
Singapore
Orders LinkContactUsLinkComponent
Drying trays, PYREX®
1500 ml, 201×201×55 mm
  CORN3175-7
 :   Corning
Drying trays, PYREX®
  CORN3175-7
 :  Corning
Drying trays, PYREX®
Drying trays, PYREX®
1500 ml, 201×201×55 mm
$109.00
1 * 4 items
These dishes made of PYREX® borosilicate glass will withstand hot air or steam sterilization and are useful for drying, staining or developing trays.
  • Excellent chemical resistance
  • Good temperature resistance
  • Easy to clean
They can be used with gel electrophoresis or paper chromatography.
 : For oven use only. Do not use over direct heat.
  • Capacity:
    1500 ml
  • Seq:
    01
  • W×D×H:
    201×201×55 mm