Order Entry
Singapore
Orders LinkContactUsLinkComponent
 

Drying trays, PYREX®

3500 ml, 272×180×50 mm
Catalog # CORN3175-10
Supplier:   Corning
Drying trays, PYREX®
Catalog # CORN3175-10
Supplier:  Corning
Drying trays, PYREX®
Drying trays, PYREX®

3500 ml, 272×180×50 mm

Price
$186.00
Log in for availability
1 * 4 items
Product Details & Documents

These dishes made of PYREX® borosilicate glass will withstand hot air or steam sterilization and are useful for drying, staining or developing trays.

  • Excellent chemical resistance
  • Good temperature resistance
  • Easy to clean

They can be used with gel electrophoresis or paper chromatography.

Caution: For oven use only. Do not use over direct heat.

 

Specifications
  • Capacity:
    3500 ml
  • Seq:
    03
  • W×D×H:
    272×180×50 mm