Order Entry
Singapore
Orders LinkContactUsLinkComponent
Separating funnels, Squibb, PYREXPLUS®

Separating funnels, Squibb, PYREXPLUS®

125 ml
Catalog # CORN66402-125
Supplier:   Corning
Separating funnels, Squibb, PYREXPLUS®
Catalog # CORN66402-125
Supplier:  Corning
Separating funnels, Squibb, PYREXPLUS®
Separating funnels, Squibb, PYREXPLUS®

125 ml

Price
$1,640.00
Log in for availability
1 * 4 items
Product Details & Documents

PYREXPLUS®, borosilicate glass.

  • Fluoropolymer resin stopcock reduces freezing problems and grease contamination
  • Coating is autoclavable to 121 °C (250 °F) moist heat or 110 °C (230 °F) dry heat
  • Resistant to thermal shock

PYREXPLUS® labware allows for a better hand grip and clamp hold. PVC coating makes cleanup faster and easier by reducing spill volume if breakage occurs. Can be written on with any standard lab marker, permanent or water-soluble. Do not use plastic-coated ware over direct or open flame.

Documents

Specifications
  • Capacity:
    125 ml
  • Seq:
    01