Order Entry
Singapore
Orders LinkContactUsLinkComponent
Separating funnels, Squibb, PYREX®
250 ml
  CORN6404-250
 :   Corning
Separating funnels, Squibb, PYREX®
  CORN6404-250
 :  Corning
Separating funnels, Squibb, PYREX®
Separating funnels, Squibb, PYREX®
250 ml
$945.00
1 * 4 items
PYREX® Borosilicate glass.
With PTFE stopcock and linear, high-density, closed-bottom polyethylene stoppers which conform to [ST] stopper dimensions.
  • Capacity:
    250 ml
  • Seq:
    03