Separating funnels, Squibb, PYREX®
1000 ml
Supplier: Corning
Jump to:
Product Details & Documents
PYREX® Borosilicate glass.
With PTFE stopcock and linear, high-density, closed-bottom polyethylene stoppers which conform to [ST] stopper dimensions.
Documents
Certificates
Search by Lot NumberSpecifications
- Capacity:1000 ml
- Seq:06