Order Entry
Singapore
Orders LinkContactUsLinkComponent
Funnels, stemless, PYREX®

Funnels, stemless, PYREX®

60 mm, 75 mm
Catalog # CORN6240-75
Supplier:   Corning
Funnels, stemless, PYREX®
Catalog # CORN6240-75
Supplier:  Corning
Funnels, stemless, PYREX®
Funnels, stemless, PYREX®

60 mm, 75 mm

Price
$1,010.00
Log in for availability
1 * 48 items
Product Details & Documents

PYREX®, borosilicate glass.

  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes

These stemless funnels are of molded construction with bottoms ground flat and have a 6 mm opening. For use in the sugar industries.

Documents

Specifications
  • Length:
    60 mm
  • Seq:
    01
  • Int.Ø top:
    75 mm